Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005249.4 | NP_005240.3 | 1 | 2263 | ||
GenBank mRNA | X74142.1 | - | 1 | 2238 |