Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGRMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006320.4 | NP_006311.2 | 3 | 1713 | ||
GenBank mRNA | AJ002030.1 | - | 3 | 680 | ||
AK091741.1 | - | 4 | 650 | |||
AK094949.1 | - | 2 | 800 | |||
AK131272.1 | - | 3 | 1713 | |||
BC016692.1 | - | 3 | 697 | 4067832 | ||
BC092478.1 | - | 3 | 728 | 4831185 |