Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301374.1 | NP_001288303.1 | 4, 5 | 712 | ||
NM_009898.3 | NP_034028.1 | 4, 5 | 625 | |||
GenBank mRNA | AF047388.1 | - | 3, 4 | 459 | ||
AF143955.1 | - | 4, 5 | 596 | |||
AF495468.1 | - | 4, 5 | 563 | |||
AK040885.1 | - | 4, 5 | 568 | |||
AK075799.1 | - | 4, 5 | 560 | |||
AK146290.1 | - | 4, 5 | 563 | |||
AK150962.1 | - | 4, 5 | 567 | |||
AK151755.1 | - | 4, 5 | 567 | |||
AK155527.1 | - | 4, 5 | 568 | |||
AK155860.1 | - | 4, 5 | 563 | |||
AK165296.1 | - | 4, 5 | 567 | |||
AK172447.1 | - | 4, 5 | 563 | |||
BC002136.1 | - | 4, 5 | 553 | 3484185 | ||
BC053398.1 | - | 4, 5 | 543 | 30076939 |