Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009921.2 | NP_034051.2 | 2 | 228 | ||
GenBank mRNA | AK158005.1 | - | 2 | 220 | ||
BC120752.1 | - | 2 | 225 | 40129675 | ||
BC120754.1 | - | 2 | 228 | 40129677 | ||
U43409.1 | - | 2 | 221 | |||
X94353.1 | - | 2 | 220 |