Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009933.4 | NP_034063.1 | 2 | 319 | ||
GenBank mRNA | AK052924.1 | - | 2 | 308 | ||
X66405.1 | - | 2 | 319 |