Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170744.1 | NP_001164215.1 | 2, 3 | 1878 | ||
NM_009980.4 | NP_034110.1 | 4, 5 | 755 | |||
GenBank mRNA | AB033123.1 | - | 4, 5 | 620 | ||
AF016508.1 | - | 2, 3 | 227 | |||
AF059735.1 | - | 2, 3 | 206 | |||
AK134399.1 | - | 3, 4 | 279 | |||
AK146713.1 | - | 4, 5 | 640 | |||
AK147871.1 | - | 4, 5 | 638 | |||
AK178844.1 | - | 4 | 431 | |||
AK180619.1 | - | 1, 2 | 110 | |||
AK196314.1 | - | 1, 2 | 110 | |||
AK205105.1 | - | 1, 2 | 110 | |||
AK205300.1 | - | 2, 3 | 263 | |||
BC013333.1 | - | 2, 3 | 262 | 4019572 | ||
BC017520.1 | - | 3, 4 | 1723 | 4511826 |