Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009983.2 | NP_034113.1 | 4, 5 | 533 | ||
GenBank mRNA | AK089294.1 | - | 4, 5 | 491 | ||
AK150363.1 | - | 4, 5 | 494 | |||
AK150364.1 | - | 4, 5 | 491 | |||
AK150579.1 | - | 4, 5 | 491 | |||
AK150622.1 | - | 4, 5 | 491 | |||
AK150636.1 | - | 4, 5 | 491 | |||
AK150694.1 | - | 4, 5 | 491 | |||
AK150768.1 | - | 4, 5 | 491 | |||
AK150946.1 | - | 4, 5 | 543 | |||
AK151256.1 | - | 4, 5 | 491 | |||
AK151275.1 | - | 4, 5 | 491 | |||
AK151276.1 | - | 4, 5 | 491 | |||
AK151374.1 | - | 4, 5 | 491 | |||
AK151829.1 | - | 4, 5 | 491 | |||
AK151962.1 | - | 4, 5 | 494 | |||
AK152036.1 | - | 4, 5 | 504 | |||
AK152045.1 | - | 4, 5 | 544 | |||
AK152168.1 | - | 4, 5 | 491 | |||
AK152187.1 | - | 4, 5 | 491 | |||
AK152188.1 | - | 4, 5 | 491 | |||
AK152406.1 | - | 4, 5 | 494 | |||
AK152458.1 | - | 4, 5 | 491 | |||
AK152567.1 | - | 4, 5 | 491 | |||
AK152578.1 | - | 4, 5 | 491 | |||
AK152593.1 | - | 4, 5 | 521 | |||
AK152638.1 | - | 4, 5 | 521 | |||
AK152680.1 | - | 4, 5 | 491 | |||
AK152828.1 | - | 4, 5 | 491 | |||
AK152881.1 | - | 4, 5 | 491 | |||
AK152892.1 | - | 4, 5 | 491 | |||
AK153191.1 | - | 4, 5 | 491 | |||
AK153291.1 | - | 4, 5 | 491 | |||
AK159097.1 | - | 4, 5 | 491 | |||
AK159209.1 | - | 4, 5 | 491 | |||
AK159440.1 | - | 4, 5 | 544 | |||
AK159490.1 | - | 4, 5 | 491 | |||
AK159512.1 | - | 4, 5 | 533 | |||
AK159580.1 | - | 4, 5 | 491 | |||
AK159642.1 | - | 2, 3 | 243 | |||
AK159743.1 | - | 4, 5 | 495 | |||
AK159896.1 | - | 4, 5 | 491 | |||
AK159970.1 | - | 4, 5 | 544 | |||
AK165227.1 | - | 4, 5 | 561 | |||
AK166864.1 | - | 4, 5 | 504 | |||
AK166938.1 | - | 4, 5 | 538 | |||
AK167143.1 | - | 4, 5 | 529 | |||
AK167366.1 | - | 4, 5 | 563 | |||
AK167698.1 | - | 4, 5 | 490 | |||
AK167750.1 | - | 4, 5 | 504 | |||
AK167867.1 | - | 4, 5 | 492 | |||
AK169139.1 | - | 4, 5 | 544 | |||
AK171195.1 | - | 4, 5 | 492 | |||
AK172583.1 | - | 4, 5 | 491 | |||
AK194256.1 | - | 1, 2 | 61 | |||
AK194597.1 | - | 1, 2 | 62 | |||
AK213259.1 | - | 1, 2 | 61 | |||
AK213518.1 | - | 1, 2 | 61 | |||
BC054758.1 | - | 4, 5 | 563 | 6413315 | ||
BC057931.1 | - | 4, 5 | 524 | 5253319 | ||
X52886.1 | - | 4, 5 | 533 | |||
X53337.1 | - | 4, 5 | 473 |