Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2d9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010006.2 | NP_034136.2 | 4, 5 | 758 | ||
GenBank mRNA | AK143963.1 | - | 4, 5 | 758 | ||
BC010593.1 | - | 4, 5 | 733 | 3964178 | ||
BC094015.1 | - | 4, 5 | 741 | 5050233 | ||
M23997.1 | - | 4, 5 | 849 | |||
M23998.1 | - | 4, 5 | 754 | |||
M27168.1 | - | 4, 5 | 733 |