Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009983.2 | NP_034113.1 | 8 | 1098 | ||
GenBank mRNA | AK089294.1 | - | 8 | 1056 | ||
AK150363.1 | - | 8 | 1059 | |||
AK150364.1 | - | 8 | 1056 | |||
AK150579.1 | - | 8 | 1056 | |||
AK150622.1 | - | 8 | 1056 | |||
AK150636.1 | - | 8 | 1056 | |||
AK150694.1 | - | 8 | 1056 | |||
AK150768.1 | - | 8 | 1056 | |||
AK150946.1 | - | 8 | 1108 | |||
AK151256.1 | - | 8 | 1056 | |||
AK151275.1 | - | 8 | 1056 | |||
AK151276.1 | - | 8 | 1056 | |||
AK151374.1 | - | 8 | 1056 | |||
AK151829.1 | - | 8 | 1056 | |||
AK151962.1 | - | 8 | 1059 | |||
AK152036.1 | - | 8 | 1069 | |||
AK152045.1 | - | 8 | 1109 | |||
AK152168.1 | - | 8 | 1056 | |||
AK152187.1 | - | 8 | 1056 | |||
AK152188.1 | - | 8 | 1056 | |||
AK152406.1 | - | 8 | 1059 | |||
AK152458.1 | - | 8 | 1056 | |||
AK152567.1 | - | 8 | 1056 | |||
AK152578.1 | - | 8 | 1056 | |||
AK152593.1 | - | 8 | 1086 | |||
AK152638.1 | - | 8 | 1068 | |||
AK152680.1 | - | 8 | 1056 | |||
AK152828.1 | - | 8 | 1056 | |||
AK152881.1 | - | 8 | 1056 | |||
AK152892.1 | - | 8 | 1056 | |||
AK153191.1 | - | 8 | 1056 | |||
AK153291.1 | - | 8 | 1056 | |||
AK159097.1 | - | 8 | 1056 | |||
AK159209.1 | - | 8 | 1056 | |||
AK159440.1 | - | 8 | 1109 | |||
AK159490.1 | - | 8 | 1057 | |||
AK159512.1 | - | 8 | 1098 | |||
AK159580.1 | - | 8 | 1056 | |||
AK159642.1 | - | 6 | 808 | |||
AK159743.1 | - | 8 | 1060 | |||
AK159896.1 | - | 8 | 1056 | |||
AK159970.1 | - | 8 | 1109 | |||
AK165227.1 | - | 8 | 1126 | |||
AK166864.1 | - | 8 | 1069 | |||
AK166938.1 | - | 8 | 1103 | |||
AK167143.1 | - | 8 | 1094 | |||
AK167366.1 | - | 8 | 1128 | |||
AK167698.1 | - | 8 | 1055 | |||
AK167750.1 | - | 8 | 1069 | |||
AK167867.1 | - | 8 | 1057 | |||
AK169139.1 | - | 8 | 1109 | |||
AK171195.1 | - | 8 | 1057 | |||
AK172583.1 | - | 8 | 1057 | |||
AK205399.1 | - | 3 | 250 | |||
AK213803.1 | - | 3 | 250 | |||
BC019682.1 | - | 4 | 453 | |||
BC048900.1 | - | 4 | 343 | |||
BC053937.1 | - | 4 | 343 | |||
BC053940.1 | - | 4 | 343 | |||
BC054758.1 | - | 8 | 1128 | 6413315 | ||
BC057931.1 | - | 8 | 1089 | 5253319 | ||
X52886.1 | - | 8 | 1098 | |||
X53337.1 | - | 8 | 1038 |