Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109750.1 | NP_001103220.1 | 2 | 446 | ||
NM_010048.3 | NP_034178.2 | 2 | 446 | |||
GenBank mRNA | AK042786.1 | - | 2 | 368 | ||
AK088131.1 | - | 2 | 376 | |||
AK129070.1 | - | 2 | 378 | |||
AK141594.1 | - | 2 | 412 | |||
AK145495.1 | - | 2 | 363 | |||
AK153026.1 | - | 2 | 234 | |||
AK161022.1 | - | 373 | ||||
AK161359.1 | - | 2 | 171 | |||
AK163872.1 | - | 2 | 408 | |||
AK163903.1 | - | 2 | 373 | |||
AK177402.1 | - | 16 | ||||
AK193194.1 | - | 1 | 16 | |||
AK215211.1 | - | 1 | 16 | |||
BC060636.1 | - | 2 | 387 | 6849298 | ||
BC062978.1 | - | 2 | 341 | 30356000 | ||
D78641.1 | - | 2 | 380 | |||
X95480.1 | - | 2 | 315 |