Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285807.1 | NP_001272736.1 | 7 | 942 | ||
NM_001285808.1 | NP_001272737.1 | 5 | 598 | |||
NM_001285810.1 | NP_001272739.1 | 5 | 598 | |||
NM_001285811.1 | NP_001272740.1 | 6 | 720 | |||
NM_001285813.1 | NP_001272742.1 | 6 | 755 | |||
NM_001285817.1 | NP_001272746.1 | 5 | 598 | |||
NM_010087.4 | NP_034217.2 | 6 | 720 | |||
NM_207650.4 | NP_997533.1 | 6 | 720 | |||
GenBank mRNA | AF143542.1 | - | 6 | 619 | ||
AF143543.1 | - | 6 | 656 | |||
AF143544.1 | - | 6 | 565 | |||
AK003829.2 | - | 6 | 640 | |||
AK019477.1 | - | 7 | 862 | |||
AK131649.1 | - | 5 | 491 | |||
AK147279.1 | - | 6 | 666 | |||
AK147359.1 | - | 5 | 555 | |||
BC040364.1 | - | 6 | 687 | 4505583 | ||
BC050765.1 | - | 5 | 506 | 6533769 | ||
X95226.1 | - | 5 | 420 | |||
X95227.1 | - | 6 | 550 |