Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eya1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252192.1 | NP_001239121.1 | 5 | 834 | ||
NM_010164.2 | NP_034294.2 | 6 | 965 | |||
GenBank mRNA | AK042412.1 | - | 5 | 757 | ||
AK153788.1 | - | 6 | 970 | |||
BC060260.1 | - | 6 | 965 | 6848408 | ||
BC066860.1 | - | 6 | 619 | 30089692 | ||
U61110.1 | - | 5 | 507 |