Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136060.2 | NP_001129532.1 | 3, 4 | 633 | ||
NM_001198886.1 | NP_001185815.1 | 3, 4 | 717 | |||
NM_010075.2 | NP_034205.1 | 3, 4 | 968 | |||
NM_207282.3 | NP_997165.2 | 3, 4 | 821 | |||
GenBank mRNA | AF092507.1 | - | 3, 4 | 976 | ||
AK031903.1 | - | 3, 4 | 594 | |||
AK051708.1 | - | 3, 4 | 799 | |||
AK082186.1 | - | 3, 4 | 892 | |||
AK158964.1 | - | 3, 4 | 822 | |||
BC048383.1 | - | 3, 4 | 484 | 6306267 | ||
BC085154.1 | - | 3, 4 | 717 | 6838230 |