Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dffa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025296.2 | NP_001020467.1 | 4 | 594 | ||
NM_010044.3 | NP_034174.2 | 4 | 594 | |||
GenBank mRNA | AB009375.1 | - | 4 | 485 | ||
AB009376.1 | - | 4 | 485 | |||
AK039227.1 | - | 4 | 591 | |||
AK051011.1 | - | 4 | 524 | |||
AK079634.1 | - | 4 | 552 | |||
AK154488.1 | - | 4 | 559 | |||
BC058213.1 | - | 4 | 539 | 5367607 | ||
BI736905.1 | - | 4 | 538 |