Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou6f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010127.3 | NP_034257.1 | 6 | 1463 | ||
GenBank mRNA | AB221558.1 | - | 5 | 753 | ||
AK006732.1 | - | 2 | 543 | |||
BC053046.1 | - | 8 | 1293 | |||
BC059830.1 | - | 6 | 1463 | 6410550 | ||
BC085139.1 | - | 11 | 1841 | 6406821 | ||
D13801.1 | - | 6 | 952 | |||
L13763.1 | - | 5 | 786 |