Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aktip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017861.1 | - | 2 | 155 | ||
AK146459.1 | - | 3 | 270 | |||
AK158704.1 | - | 3 | 272 | |||
AK209938.1 | - | 2 | 85 | |||
BC008130.1 | - | 4 | 275 | 3498984 | ||
BC034861.1 | - | 3 | 279 | |||
CJ153808.1 | - | 4 | 331 | |||
X71978.1 | - | 3 | 294 | |||
Z67963.1 | - | 3 | 294 |