Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010176.4 | NP_034306.2 | 14 | 1551 | ||
GenBank mRNA | AK143759.1 | - | 14 | 1380 | ||
AK158808.1 | - | 14 | 1437 | |||
AK177342.1 | - | 451 | ||||
AK215090.1 | - | 1 | 67 | |||
BC010767.1 | - | 14 | 1336 | 4242421 | ||
M84145.1 | - | 14 | 1377 | |||
Z11774.1 | - | 14 | 1363 |