Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010231.2 | NP_034361.1 | 7 | 1236 | ||
GenBank mRNA | AK034946.1 | - | 1 | 879 | ||
BC011229.1 | - | 7 | 1236 | 4196446 | ||
D16215.1 | - | 7 | 1126 | |||
U87456.1 | - | 7 | 1221 |