Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159544.1 | NP_001153016.1 | 6, 7 | 1774 | ||
NM_010237.3 | NP_034367.2 | 7, 8 | 1618 | |||
GenBank mRNA | AK028855.1 | - | 7, 8 | 1558 | ||
AK052614.1 | - | 7, 8 | 1490 | |||
AK085191.1 | - | 6, 7 | 1619 | |||
AK166379.1 | - | 7, 8 | 1618 | |||
BC007137.1 | - | 7, 8 | 1631 | 3601246 | ||
L36132.1 | - | 6, 7 | 1653 | |||
Z48757.1 | - | 7, 8 | 1483 |