Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010294.1 | NP_034424.1 | 1 | 159 | ||
GenBank mRNA | AF117734.1 | - | 1 | 159 | ||
AK075761.1 | - | 1 | 72 | |||
BC050764.1 | - | 1 | 172 | 6774427 | ||
BC061147.1 | - | 1 | 138 | 6770827 |