Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010350.2 | NP_034480.2 | 5 | 1246 | ||
GenBank mRNA | AK147390.1 | - | 4 | 974 | ||
AK165770.1 | - | 2 | 101 | |||
BC137849.1 | - | 3 | 636 | 8860868 | ||
D10694.1 | - | 2 | 473 |