Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172117.1 | NP_001165588.1 | 3, 4 | 455 | ||
NM_010407.4 | NP_034537.2 | 3, 4 | 455 | |||
GenBank mRNA | AK149736.1 | - | 3, 4 | 421 | ||
AK150290.1 | - | 3, 4 | 429 | |||
AK150709.1 | - | 3, 4 | 438 | |||
AK152506.1 | - | 3, 4 | 429 | |||
AK155975.1 | - | 3, 4 | 368 | |||
AK159234.1 | - | 3, 4 | 387 | |||
AK165315.1 | - | 3, 4 | 420 | |||
BC010478.1 | - | 3, 4 | 451 | 3978518 | ||
J03023.1 | - | 3, 4 | 344 | |||
Y00487.1 | - | 3, 4 | 350 |