Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcrt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010410.2 | NP_034540.1 | 2 | 486 | ||
GenBank mRNA | AF019566.1 | - | 2 | 487 | ||
AF041242.1 | - | 2 | 486 | |||
BC116943.1 | - | 2 | 479 | 40126262 | ||
BC116969.1 | - | 2 | 479 | 40126288 |