Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF065145.1 | - | 2 | 852 | ||
AK132272.1 | - | 2 | 953 | |||
BC137982.1 | - | 2 | 849 | 8861001 |