Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159551.1 | NP_001153023.1 | 5, 6 | 665 | ||
NM_010376.4 | NP_034506.1 | 5, 6 | 665 | |||
GenBank mRNA | AF483214.1 | - | 5, 6 | 632 | ||
AF515662.1 | - | 5, 6 | 544 | |||
AJ345032.1 | - | 5, 6 | 675 | |||
AK007664.1 | - | 5, 6 | 647 | |||
AK012600.1 | - | 5, 6 | 658 | |||
AK028109.1 | - | 5, 6 | 634 | |||
AK049101.1 | - | 5, 6 | 639 | |||
AK050664.1 | - | 5, 6 | 608 | |||
AK080966.1 | - | 5, 6 | 627 | |||
AK159176.1 | - | 5, 6 | 639 | |||
AK169922.1 | - | 5, 6 | 622 | |||
BC045195.1 | - | 1, 2 | 21 | |||
DQ168449.1 | - | 5, 6 | 681 |