Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290822.1 | NP_001277751.1 | 5 | 1584 | ||
NM_010329.3 | NP_034459.2 | 6 | 1623 | |||
GenBank mRNA | AK158855.1 | - | 6 | 1623 | ||
BC026551.1 | - | 6 | 1505 | 4985288 | ||
M73748.1 | - | 6 | 1477 |