Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010345.4 | NP_034475.2 | 5, 6 | 855 | ||
GenBank mRNA | AF022072.1 | - | 5, 6 | 829 | ||
AK030727.1 | - | 5, 6 | 790 | |||
AK032927.1 | - | 5, 6 | 836 | |||
AK080969.1 | - | 1, 2 | 1289 | |||
AK163841.1 | - | 5, 6 | 836 | |||
BC053842.1 | - | 5, 6 | 822 |