Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010361.2 | NP_034491.2 | 5 | 1260 | ||
GenBank mRNA | AK002392.1 | - | 5 | 790 | ||
AK158939.1 | - | 1251 | ||||
BC012707.1 | - | 5 | 818 | 3994154 | ||
U48420.1 | - | 5 | 796 | |||
X98056.1 | - | 5 | 665 |