Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278623.1 | NP_001265552.1 | 20 | 2450 | ||
NM_001278624.1 | NP_001265553.1 | 20 | 2588 | |||
NM_152995.5 | NP_694540.3 | 20 | 2490 | |||
NR_103795.1 | 21 | 2630 | ||||
GenBank mRNA | AB085695.1 | - | 20 | 2446 | ||
AB181916.1 | - | 20 | 2570 | |||
AK131285.1 | - | 18 | 2036 | |||
AK302440.1 | - | 20 | 2448 | |||
AY134856.1 | - | 4 | 494 | |||
BC018019.1 | - | 3 | 185 | |||
BC051193.1 | - | 21 | 2601 | 5271401 |