Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048061.1 | NP_001041526.1 | 2, 3 | 462 | ||
NM_010448.3 | NP_034578.1 | 2, 3 | 462 | |||
GenBank mRNA | AK013384.1 | - | 2, 3 | 429 | ||
AK013709.1 | - | 428 | ||||
AK076132.1 | - | 2, 3 | 425 | |||
AK165609.1 | - | 3, 4 | 409 | |||
BC043069.1 | - | 2, 3 | 416 | 6402930 | ||
CA325334.1 | - | 1, 2 | 160 | |||
D90151.1 | - | 2, 3 | 417 | |||
DQ397821.1 | - | 1, 2 | 224 | |||
DQ397822.1 | - | 1, 2 | 224 | |||
L36663.1 | - | 421 |