Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010476.3 | NP_034606.3 | 2, 3 | 306 | ||
GenBank mRNA | AK028380.1 | - | 2, 3 | 262 | ||
AK050211.1 | - | 2, 3 | 263 | |||
AK077914.1 | - | 2, 3 | 304 | |||
BC011464.1 | - | 2, 3 | 270 | |||
BY228199.1 | - | 2, 3 | 312 | |||
Y15733.1 | - | 2, 3 | 296 |