Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010477.4 | NP_034607.3 | 2 | 291 | ||
GenBank mRNA | AK088844.1 | - | 2 | 205 | ||
AK143882.1 | - | 2 | 196 | |||
AK146831.1 | - | 2 | 243 | |||
AK218891.1 | - | 2 | 184 | |||
AK219639.1 | - | 1 | 35 | |||
BC016400.1 | - | 2 | 177 | 3585270 | ||
BC106112.1 | - | 2 | 192 | |||
BY014148.1 | - | 2 | 300 | |||
X55023.1 | - | 2 | 153 |