Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010449.4 | NP_034579.3 | 1, 2 | 759 | ||
GenBank mRNA | AK083575.1 | - | 1, 2 | 760 | ||
AK143943.1 | - | 2, 3 | 551 | |||
BC138097.1 | - | 1, 2 | 693 | 8861116 | ||
BC138098.1 | - | 1, 2 | 696 | 8861117 | ||
M22115.1 | - | 1, 2 | 728 |