Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010477.4 | NP_034607.3 | 2, 3 | 338 | ||
GenBank mRNA | AK088844.1 | - | 2, 3 | 252 | ||
AK143882.1 | - | 2, 3 | 243 | |||
AK146831.1 | - | 2, 3 | 290 | |||
AK218891.1 | - | 2, 3 | 231 | |||
AK219639.1 | - | 1, 2 | 82 | |||
BC016400.1 | - | 2, 3 | 224 | 3585270 | ||
BC106112.1 | - | 2, 3 | 239 | |||
X55023.1 | - | 2, 3 | 200 |