Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010556.4 | NP_034686.2 | 2, 3 | 230 | ||
GenBank mRNA | A02046.1 | - | 2, 3 | 228 | ||
AK153634.1 | - | 2, 3 | 231 | |||
BC125552.1 | - | 2, 3 | 225 | 40130067 | ||
BC125554.1 | - | 2, 3 | 225 | 40130069 | ||
DQ788721.1 | - | 2, 3 | 201 | |||
K01668.1 | - | 2, 3 | 228 | |||
K01850.1 | - | 2, 3 | 228 |