Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010468.2 | NP_034598.2 | 3 | 1979 | ||
GenBank mRNA | AK133705.1 | - | 3 | 1847 | ||
AK138525.1 | - | 3 | 1980 | |||
AK140904.1 | - | 1 | 1627 | |||
U56400.1 | - | 2 | 1655 | |||
X73573.1 | - | 2 | 1635 |