Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010469.2 | NP_034599.2 | 6 | 2227 | ||
GenBank mRNA | AK135479.1 | - | 2 | 1018 | ||
AK136751.1 | - | 1025 | ||||
BC139206.1 | - | 2 | 1119 | 8862228 | ||
BC139207.1 | - | 2 | 1119 | 8862229 | ||
J03770.1 | - | 2 | 1198 |