Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elavl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010485.3 | NP_034615.2 | 6 | 1392 | ||
GenBank mRNA | AK011870.1 | - | 6 | 1341 | ||
AK028769.1 | - | 1 | 76 | |||
AK034536.1 | - | 6 | 1380 | |||
AK078333.1 | - | 6 | 1708 | |||
AK080365.1 | - | 6 | 1350 | |||
AK089088.1 | - | 5 | 1099 | |||
AK148528.1 | - | 6 | 1320 | |||
AK161667.1 | - | 5 | 1109 | |||
AK205357.1 | - | 1 | 243 | |||
AK212808.1 | - | 1 | 235 |