Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111274.1 | NP_001104744.1 | 5 | 775 | ||
NM_001111275.1 | NP_001104745.1 | 4 | 805 | |||
NM_001111276.1 | NP_001104746.1 | 4 | 723 | |||
NM_010512.4 | NP_034642.2 | 5 | 857 | |||
GenBank mRNA | AK081019.1 | - | 4 | 794 | ||
AK155435.1 | - | 5 | 1277 | |||
AK165471.1 | - | 4 | 695 | |||
AY878192.1 | - | 4 | 603 | |||
AY878193.1 | - | 5 | 655 | |||
BC012409.1 | - | 4 | 512 | 4194295 | ||
X04480.1 | - | 4 | 585 | |||
X04482.1 | - | 5 | 557 |