Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Laptm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010686.3 | NP_034816.1 | 6, 7 | 629 | ||
GenBank mRNA | AF364050.1 | - | 6, 7 | 647 | ||
AK031802.1 | - | 6, 7 | 643 | |||
AK050633.1 | - | 6, 7 | 644 | |||
AK089432.1 | - | 6, 7 | 679 | |||
AK089508.1 | - | 6, 7 | 652 | |||
AK089535.1 | - | 6, 7 | 644 | |||
AK150299.1 | - | 6, 7 | 640 | |||
AK152918.1 | - | 6, 7 | 640 | |||
AK153877.1 | - | 6, 7 | 643 | |||
AK155353.1 | - | 1, 2 | 86 | |||
AK155744.1 | - | 6, 7 | 637 | |||
AK163146.1 | - | 6, 7 | 644 | |||
AK170438.1 | - | 6, 7 | 631 | |||
AK170690.1 | - | 6, 7 | 663 | |||
AK170867.1 | - | 6, 7 | 647 | |||
AK171053.1 | - | 6, 7 | 654 | |||
AK171057.1 | - | 6, 7 | 679 | |||
AK171105.1 | - | 6, 7 | 644 | |||
AK171116.1 | - | 6, 7 | 644 | |||
AK171149.1 | - | 6, 7 | 652 | |||
AK171172.1 | - | 644 | ||||
AK171193.1 | - | 6, 7 | 644 | |||
AK171213.1 | - | 6, 7 | 644 | |||
AK171236.1 | - | 6, 7 | 644 | |||
AK171244.1 | - | 6, 7 | 644 | |||
AK171250.1 | - | 6, 7 | 665 | |||
AK171269.1 | - | 6, 7 | 681 | |||
AK171270.1 | - | 6, 7 | 647 | |||
AK171274.1 | - | 6, 7 | 637 | |||
AK171280.1 | - | 6, 7 | 677 | |||
AK171285.1 | - | 6, 7 | 681 | |||
AK171481.1 | - | 6, 7 | 680 | |||
AK171547.1 | - | 6, 7 | 644 | |||
AK171562.1 | - | 6, 7 | 635 | |||
AK171571.1 | - | 6, 7 | 661 | |||
AK171575.1 | - | 6, 7 | 679 | |||
AK172306.1 | - | 6, 7 | 644 | |||
AK172486.1 | - | 6, 7 | 679 | |||
AK172707.1 | - | 6, 7 | 644 | |||
AK172717.1 | - | 3, 4 | 347 | |||
AK172885.1 | - | 6, 7 | 619 | |||
BC020993.1 | - | 6, 7 | 629 | 4037686 | ||
BC055057.1 | - | 6, 7 | 616 | 6529760 | ||
BC114403.1 | - | 6, 7 | 609 | |||
BC158032.1 | - | 6, 7 | 825 | 9007335 | ||
U29539.1 | - | 6, 7 | 647 | |||
U51239.1 | - | 6, 7 | 668 |