Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010696.3 | NP_034826.2 | 1, 2 | 235 | ||
GenBank mRNA | AK036727.1 | - | 1, 2 | 293 | ||
AK037970.1 | - | 1, 2 | 269 | |||
AK088552.1 | - | 1, 2 | 357 | |||
AK168134.1 | - | 1, 2 | 251 | |||
AK170491.1 | - | 1, 2 | 512 | |||
BC006948.1 | - | 1, 2 | 235 | 3603181 | ||
U20159.1 | - | 1, 2 | 274 |