Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013954.1 | - | 3 | 516 | ||
AK133957.1 | - | 3 | 246 | |||
BC040362.1 | - | 3 | 307 | 4506177 | ||
BC049100.1 | - | 3 | 505 | 5698182 | ||
D50366.1 | - | 3 | 420 | |||
D50367.1 | - | 3 | 420 |