Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010587.2 | NP_034717.2 | 30 | 4079 | ||
GenBank mRNA | AF132481.1 | - | 29 | 3818 | ||
AK082606.1 | - | 9 | 942 | |||
AY257970.1 | - | 2 | 132 |