Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB000494.1 | - | 5 | 859 | ||
AB000495.1 | - | 5 | 859 | |||
AB000496.1 | - | 5 | 859 | |||
AB000497.1 | - | 5 | 859 | |||
AB000498.1 | - | 5 | 859 | |||
AB000499.1 | - | 5 | 859 | |||
AB000500.1 | - | 5 | 859 | |||
AB000501.1 | - | 5 | 859 | |||
AB000502.1 | - | 5 | 859 | |||
AB000503.1 | - | 5 | 859 | |||
AB000504.1 | - | 5 | 859 | |||
AF490773.1 | - | 5 | 773 | |||
AK135059.1 | - | 4 | 431 | |||
AK139411.1 | - | 5 | 604 | |||
AK161558.1 | - | 3 | 2198 | |||
BC145453.1 | - | 5 | 903 | 9054011 |