Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK141813.1 | - | 2 | 280 | ||
AK164484.1 | - | 2 | 281 | |||
BC063768.1 | - | 2 | 262 | |||
CJ066818.1 | - | 2 | 297 | |||
CV563459.1 | - | 1 | 39 | |||
U14166.1 | - | 2 | 303 | |||
U15159.1 | - | 2 | 278 |