Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcpt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010779.2 | NP_034909.2 | 4 | 533 | ||
GenBank mRNA | AB051900.1 | - | 4 | 481 | ||
AK144385.1 | - | 4 | 534 | |||
AY007569.1 | - | 4 | 533 | |||
M55617.1 | - | 533 | ||||
X68804.1 | - | 4 | 480 |