Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010739.2 | NP_034869.1 | 11, 12 | 1728 | ||
GenBank mRNA | AK132274.1 | - | 10, 11 | 1573 | ||
BC024321.1 | - | 10, 11 | 1694 | 4219376 | ||
J04634.1 | - | 11, 12 | 1728 |