Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccndbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010761.2 | NP_034891.2 | 11 | 1471 | ||
GenBank mRNA | AK154285.1 | - | 11 | 1425 | ||
AK163150.1 | - | 11 | 1384 | |||
AK167345.1 | - | 11 | 1437 | |||
AK211891.1 | - | 1 | 209 | |||
BB517577.1 | - | 1 | 179 | |||
BC001984.1 | - | 11 | 1407 | 3489973 | ||
BC004673.1 | - | 5 | 891 | |||
BC005802.1 | - | 10 | 1480 | 3707919 | ||
U50734.1 | - | 11 | 1392 |