Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010829.2 | NP_034959.2 | 2 | 330 | ||
GenBank mRNA | AK011426.1 | - | 2 | 314 | ||
AK078467.1 | - | 2 | 281 | |||
AK083388.1 | - | 2 | 326 | |||
AK133907.1 | - | 2 | 323 | |||
AK161089.1 | - | 2 | 324 | |||
BC150759.1 | - | 2 | 249 | 9087670 | ||
BY710130.1 | - | 2 | 312 | |||
M24919.1 | - | 2 | 613 | |||
M80360.1 | - | 2 | 327 |