Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177352.1 | NP_001170823.1 | 2 | 715 | ||
NM_001177353.1 | NP_001170824.1 | 2 | 712 | |||
NM_001177354.1 | NP_001170825.1 | 2 | 712 | |||
NM_010849.4 | NP_034979.3 | 2 | 715 | |||
GenBank mRNA | AK087961.1 | - | 2 | 507 | ||
AK133952.1 | - | 2 | 489 | |||
AK145084.1 | - | 2 | 669 | |||
BC006728.1 | - | 1 | 189 | |||
BC138931.1 | - | 2 | 178 | 8861953 | ||
BC138932.1 | - | 2 | 178 | 8861954 | ||
X01023.1 | - | 2 | 695 |